Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP5PI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333086.1 | NP_001322580.1 | 5 - 6 | 476 | 94 | |
NM_001333087.1 | NP_001322581.1 | 1 - 2 | 248 | 94 | ||
NM_103135.4 | NP_564437.1 | 5 - 6 | 537 | 94 | ||
NM_179414.1 | NP_849745.1 | 5 - 6 | 471 | 94 | ||
NM_202231.1 | NP_973960.1 | 5 - 6 | 471 | 94 | ||
GenBank mRNA | AK227368.1 | - | 5 - 6 | 427 | 94 | |
BT006448.1 | - | 5 - 6 | 406 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |