Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333091.1 | NP_001320467.1 | 5 - 6 | 829 | 80 | |
NM_103144.5 | NP_174683.1 | 4 - 5 | 963 | 80 | ||
GenBank mRNA | AK229715.1 | - | 4 - 5 | 857 | 80 | |
FJ708645.1 | - | 4 - 5 | 449 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |