Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103188.3 | NP_174724.1 | 2 - 3 | 226 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |