Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103188.3 | NP_174724.1 | 9 - 10 | 1784 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |