Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333179.1 | NP_001319154.1 | 2 - 3 | 254 | 134 | |
NM_103322.3 | NP_564472.1 | 2 - 3 | 297 | 134 | ||
GenBank mRNA | BT015146.1 | - | 2 - 3 | 245 | 134 | |
BT015658.1 | - | 2 - 3 | 217 | 134 | ||
DQ027044.1 | - | 2 - 3 | 217 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |