Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103408.3 | NP_564479.1 | 7 - 8 | 1576 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |