Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036069.1 | NP_001031146.1 | 5 - 6 | 945 | 91 | |
NM_001084202.1 | NP_001077671.1 | 3 - 4 | 771 | 91 | ||
NM_001198219.1 | NP_001185148.1 | 5 - 6 | 1061 | 91 | ||
NM_001198220.1 | NP_001185149.1 | 5 - 6 | 1064 | 91 | ||
NM_001198221.1 | NP_001185150.1 | 5 - 6 | 1070 | 91 | ||
NM_001198222.1 | NP_001185151.1 | 5 - 6 | 1067 | 91 | ||
NM_001198223.1 | NP_001185152.1 | 5 - 6 | 1073 | 91 | ||
NM_103469.4 | NP_175009.1 | 5 - 6 | 1161 | 91 | ||
NM_202237.1 | NP_973966.1 | 5 - 6 | 947 | 91 | ||
GenBank mRNA | AF327421.1 | - | 5 - 6 | 970 | 91 | |
AF361101.1 | - | 4 - 5 | 907 | 91 | ||
AF361809.1 | - | 5 - 6 | 966 | 91 | ||
AX765804.1 | - | 4 - 5 | 907 | 91 | ||
AY039544.1 | - | 5 - 6 | 970 | 91 | ||
AY052743.1 | - | 4 - 5 | 907 | 91 | ||
AY057487.1 | - | 5 - 6 | 964 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |