Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084207.1 | NP_001077676.1 | 6 - 7 | 718 | 132 | |
NM_103541.3 | NP_175081.1 | 7 - 8 | 870 | 132 | ||
NM_179439.4 | NP_849770.1 | 6 - 7 | 1082 | 132 | ||
GenBank mRNA | AY072122.1 | - | 6 - 7 | 984 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |