Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103546.4 | NP_175086.1 | 4 - 5 | 1392 | 105 | |
GenBank mRNA | AK229399.1 | - | 4 - 5 | 1117 | 105 | |
AY072322.1 | - | 4 - 5 | 1116 | 105 | ||
AY074380.1 | - | 4 - 5 | 1051 | 105 | ||
AY117292.1 | - | 4 - 5 | 1031 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Arabidopsis thaliana Probe spans exons |