Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPQ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333231.1 | NP_001319163.1 | 2 - 3 | 396 | 62 | |
NM_103552.4 | NP_175092.1 | 1 - 2 | 450 | 62 | ||
NM_202242.2 | NP_973971.1 | 1 - 2 | 265 | 62 | ||
GenBank mRNA | AF134131.1 | - | 1 - 2 | 233 | 62 | |
AK221702.1 | - | 1 - 2 | 243 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |