Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPQ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333231.1 | NP_001319163.1 | 3 - 4 | 507 | 56 | |
NM_103552.4 | NP_175092.1 | 2 - 3 | 561 | 56 | ||
NM_202242.2 | NP_973971.1 | 2 - 3 | 376 | 56 | ||
GenBank mRNA | AF134131.1 | - | 2 - 3 | 344 | 56 | |
AK221702.1 | - | 2 - 3 | 354 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |