Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATUBC2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333242.1 | NP_001323221.1 | 2 - 3 | 263 | 135 | |
NM_103582.4 | NP_564493.1 | 2 - 3 | 263 | 135 | ||
GenBank mRNA | AK176757.1 | - | 2 - 3 | 193 | 135 | |
BT026105.1 | - | 2 - 3 | 137 | 135 | ||
DQ027029.1 | - | 2 - 3 | 137 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |