Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCA3;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103617.4 | NP_175156.1 | 3 - 4 | 891 | 97 | |
NM_202254.2 | NP_973983.1 | 3 - 4 | 891 | 97 | ||
GenBank mRNA | AK226253.1 | - | 3 - 4 | 897 | 97 | |
AY087628.1 | - | 3 - 4 | 873 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |