Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRAMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103618.3 | NP_175157.1 | 2 - 3 | 907 | 80 | |
GenBank mRNA | AF410320.1 | - | 2 - 3 | 799 | 80 | |
AY102128.1 | - | 2 - 3 | 710 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |