Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103665.3 | NP_175203.2 | 2 - 3 | 506 | 78 | |
GenBank mRNA | AK119114.1 | - | 2 - 3 | 455 | 78 | |
BT005269.1 | - | 2 - 3 | 431 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |