Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KU80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333292.1 | NP_001322094.1 | 7 - 8 | 1249 | 69 | |
NM_001333293.1 | NP_001322093.1 | 6 - 7 | 1177 | 69 | ||
NM_103701.2 | NP_564520.1 | 7 - 8 | 1297 | 69 | ||
GenBank mRNA | AF283758.1 | - | 7 - 8 | 1227 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |