Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123974.1 | NP_001117446.1 | 2 - 3 | 851 | 158 | |
NM_103707.5 | NP_175245.1 | 3 - 4 | 450 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |