Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333320.1 | NP_001321802.1 | 2 - 3 | 418 | 86 | |
NM_103724.2 | NP_175261.1 | 2 - 3 | 418 | 86 | ||
GenBank mRNA | AK228479.1 | - | 2 - 3 | 271 | 86 | |
U95143.1 | - | 2 - 3 | 414 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |