Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333320.1 | NP_001321802.1 | 4 - 5 | 689 | 66 | |
NM_103724.2 | NP_175261.1 | 4 - 5 | 689 | 66 | ||
GenBank mRNA | AK228479.1 | - | 4 - 5 | 542 | 66 | |
U95143.1 | - | 4 - 5 | 685 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |