Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHR18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333322.1 | NP_001321270.1 | 6 - 7 | 671 | 74 | |
NM_001333323.1 | NP_001321271.1 | 6 - 7 | 548 | 74 | ||
NM_001333324.1 | NP_001321269.1 | 6 - 7 | 650 | 74 | ||
NM_103728.5 | NP_175265.3 | 6 - 7 | 656 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |