Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YSL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333329.1 | NP_001321378.1 | 4 - 5 | 722 | 99 | |
NM_103733.4 | NP_564525.1 | 5 - 6 | 1162 | 99 | ||
GenBank mRNA | AY039600.1 | - | 5 - 6 | 1082 | 99 | |
BT010472.1 | - | 5 - 6 | 1038 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |