Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HON5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103758.3 | NP_175295.1 | 1 - 2 | 602 | 81 | |
GenBank mRNA | BT023425.1 | - | 1 - 2 | 289 | 81 | |
BT025318.1 | - | 1 - 2 | 240 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |