Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZIP61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339908.1 | NP_001327593.1 | 1 - 2 | 923 | 93 | |
NM_115674.4 | NP_191371.1 | 1 - 2 | 1066 | 93 | ||
GenBank mRNA | AF401300.1 | - | 1 - 2 | 626 | 93 | |
AY087356.1 | - | 1 - 2 | 846 | 93 | ||
AY093053.1 | - | 1 - 2 | 820 | 93 | ||
BT008728.1 | - | 1 - 2 | 626 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |