Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339909.1 | NP_001319790.1 | 25 - 26 | 4021 | 75 | |
NM_001339911.1 | NP_001327764.1 | 25 - 26 | 4192 | 75 | ||
NM_115678.2 | NP_191375.1 | 24 - 25 | 3670 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |