Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115730.4 | NP_191427.1 | 3 - 4 | 430 | 103 | |
GenBank mRNA | AF324717.3 | - | 3 - 4 | 403 | 103 | |
AF326909.1 | - | 3 - 4 | 401 | 103 | ||
AF339728.1 | - | 3 - 4 | 310 | 103 | ||
AY042850.1 | - | 3 - 4 | 401 | 103 | ||
AY081487.1 | - | 3 - 4 | 310 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |