Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115736.2 | NP_191433.1 | 7 - 8 | 971 | 61 | |
GenBank mRNA | AY099611.1 | - | 7 - 8 | 971 | 61 | |
BT002169.1 | - | 7 - 8 | 907 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |