Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084842.1 | NP_001078311.1 | 5 - 6 | 772 | 103 | |
NM_001203201.2 | NP_001190130.1 | 7 - 8 | 1122 | 103 | ||
NM_001339938.1 | NP_001325710.1 | 4 - 5 | 739 | 103 | ||
NM_115740.3 | NP_191437.1 | 5 - 6 | 1093 | 103 | ||
GenBank mRNA | AY086196.1 | - | 5 - 6 | 793 | 103 | |
DQ446777.1 | - | 4 - 5 | 451 | 103 | ||
JQ973091.1 | - | 4 - 5 | 472 | 103 | ||
M55550.1 | - | 4 - 5 | 478 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |