Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115776.4 | NP_191473.2 | 2 - 3 | 2632 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |