Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115776.4 | NP_191473.2 | 4 - 5 | 3298 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |