Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203206.1 | NP_001190135.1 | 14 - 15 | 2278 | 137 | |
NM_001339969.1 | NP_001326041.1 | 14 - 15 | 3145 | 137 | ||
NM_001339970.1 | NP_001326042.1 | 13 - 14 | 2287 | 137 | ||
NM_001339971.1 | NP_001326040.1 | 13 - 14 | 2058 | 137 | ||
NM_115803.3 | NP_191500.2 | 13 - 14 | 2177 | 137 | ||
GenBank mRNA | AJ459823.1 | - | 13 - 14 | 1825 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |