Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203206.1 | NP_001190135.1 | 7 - 8 | 1331 | 93 | |
NM_001339969.1 | NP_001326041.1 | 7 - 8 | 2198 | 93 | ||
NM_001339971.1 | NP_001326040.1 | 6 - 7 | 1111 | 93 | ||
NM_115803.3 | NP_191500.2 | 6 - 7 | 1230 | 93 | ||
GenBank mRNA | AJ459823.1 | - | 6 - 7 | 878 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |