Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035819.1 | NP_001030896.1 | 8 - 9 | 3176 | 75 | |
NM_001203209.1 | NP_001190138.1 | 9 - 10 | 3103 | 75 | ||
NM_001339998.1 | NP_001326481.1 | 9 - 10 | 3171 | 75 | ||
NM_115839.4 | NP_191536.2 | 9 - 10 | 3142 | 75 | ||
GenBank mRNA | AY227252.1 | - | 8 - 9 | 2936 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |