Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSY5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340022.1 | NP_001327513.1 | 18 - 19 | 1477 | 127 | |
NM_115873.3 | NP_191569.2 | 18 - 19 | 1462 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |