Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSY5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340022.1 | NP_001327513.1 | 9 - 10 | 755 | 74 | |
NM_001340023.1 | NP_001327512.1 | 9 - 10 | 755 | 74 | ||
NM_115873.3 | NP_191569.2 | 9 - 10 | 740 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |