Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203212.1 | NP_001190141.1 | 14 - 15 | 1881 | 87 | |
NM_001340039.1 | NP_001327362.1 | 14 - 15 | 1874 | 87 | ||
NM_001340040.1 | NP_001327363.1 | 13 - 14 | 1908 | 87 | ||
NM_115897.5 | NP_191592.5 | 13 - 14 | 1879 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |