Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203212.1 | NP_001190141.1 | 6 - 7 | 735 | 102 | |
NM_115897.5 | NP_191592.5 | 5 - 6 | 733 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |