Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115928.5 | NP_191623.1 | 3 - 4 | 266 | 144 | |
GenBank mRNA | AB073181.1 | - | 3 - 4 | 153 | 144 | |
AY062844.1 | - | 3 - 4 | 176 | 144 | ||
BT001196.1 | - | 2 - 3 | 92 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |