Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115928.5 | NP_191623.1 | 5 - 6 | 441 | 108 | |
GenBank mRNA | AB073181.1 | - | 5 - 6 | 328 | 108 | |
AY062844.1 | - | 5 - 6 | 351 | 108 | ||
AY088249.1 | - | 5 - 6 | 351 | 108 | ||
BT001196.1 | - | 4 - 5 | 267 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |