Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115947.3 | NP_567105.1 | 1 - 2 | 622 | 129 | |
GenBank mRNA | AF507915.1 | - | 1 - 2 | 307 | 129 | |
AY087739.1 | - | 1 - 2 | 510 | 129 | ||
AY099661.1 | - | 1 - 2 | 532 | 129 | ||
AY128849.1 | - | 1 - 2 | 307 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |