Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115977.4 | NP_191672.1 | 3 - 4 | 578 | 86 | |
GenBank mRNA | DQ370437.1 | - | 2 - 3 | 176 | 86 | |
KJ138782.1 | - | 2 - 3 | 176 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |