Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115977.4 | NP_191672.1 | 5 - 6 | 772 | 125 | |
GenBank mRNA | DQ370437.1 | - | 4 - 5 | 370 | 125 | |
KJ138782.1 | - | 4 - 5 | 370 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |