Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115978.4 | NP_567109.1 | 5 - 6 | 1318 | 76 | |
GenBank mRNA | AF360295.1 | - | 5 - 6 | 1204 | 76 | |
AF395057.1 | - | 5 - 6 | 1150 | 76 | ||
AY051056.1 | - | 5 - 6 | 1150 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |