Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115979.5 | NP_191674.1 | 4 - 5 | 1620 | 109 | |
GenBank mRNA | AY050866.1 | - | 4 - 5 | 1341 | 109 | |
AY096757.1 | - | 4 - 5 | 1182 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |