Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128553.3 | NP_180560.1 | 1 - 2 | 639 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |