Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202710.1 | NP_001189639.1 | 9 - 10 | 1474 | 56 | |
NM_128592.3 | NP_180598.1 | 9 - 10 | 1478 | 56 | ||
GenBank mRNA | BT000465.1 | - | 9 - 10 | 1421 | 56 | |
BT008877.1 | - | 9 - 10 | 1315 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |