Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202711.2 | NP_001189640.1 | 3 - 4 | 353 | 68 | |
NM_001336255.1 | NP_001318319.1 | 3 - 4 | 489 | 68 | ||
NM_128594.4 | NP_565699.1 | 2 - 3 | 344 | 68 | ||
GenBank mRNA | AF486848.1 | - | 1 - 2 | 150 | 68 | |
AY050952.1 | - | 2 - 3 | 286 | 68 | ||
AY085814.1 | - | 2 - 3 | 266 | 68 | ||
AY091344.1 | - | 1 - 2 | 150 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |