Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202711.2 | NP_001189640.1 | 4 - 5 | 446 | 61 | |
NM_001336255.1 | NP_001318319.1 | 4 - 5 | 582 | 61 | ||
NM_128594.4 | NP_565699.1 | 3 - 4 | 437 | 61 | ||
GenBank mRNA | AF486848.1 | - | 2 - 3 | 243 | 61 | |
AY050952.1 | - | 3 - 4 | 379 | 61 | ||
AY085814.1 | - | 3 - 4 | 359 | 61 | ||
AY091344.1 | - | 2 - 3 | 243 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |