Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161240.2 | NP_001154712.2 | 14 - 15 | 1638 | 69 | |
NM_121303.8 | NP_196804.6 | 14 - 15 | 1700 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |