Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121342.4 | NP_196843.1 | 2 - 3 | 1905 | 71 | |
GenBank mRNA | AB158416.1 | - | 2 - 3 | 1862 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |