Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121349.4 | NP_196850.1 | 3 - 4 | 699 | 58 | |
GenBank mRNA | AY054652.1 | - | 3 - 4 | 593 | 58 | |
AY086149.1 | - | 3 - 4 | 548 | 58 | ||
AY128736.1 | - | 2 - 3 | 371 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |